Assay Details
| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245747.1 | XP_021101406.1 | 2 - 3 | 405 | 103 | |
| XM_021245749.1 | XP_021101408.1 | 2 - 3 | 405 | 103 | ||
More Information
Set Membership: |
|

Il11
Genomic Map