Assay Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075201.2 | XP_012930655.1 | 12 - 13 | 1687 | 59 | |
| XM_021265060.1 | XP_021120719.1 | 12 - 13 | 2239 | 59 | ||
| XM_021265061.1 | XP_021120720.1 | 13 - 14 | 1752 | 59 | ||
More Information
Set Membership: |
|

Fam149a
Genomic Map