Assay Details
| Gene Symbol: Ccz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840464.2 | XP_004840521.1 | 11 - 12 | 1105 | 66 | |
| XM_013070632.2 | XP_012926086.1 | 10 - 11 | 1021 | 66 | ||
More Information
Set Membership: |
|

Ccz1
Genomic Map