Assay Details
Gene Symbol: Pts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856588.2 | XP_004856645.2 | 4 - 5 | 446 | 88 | |
XM_013076670.2 | XP_012932124.2 | 4 - 5 | 360 | 88 | ||
XM_021237743.1 | XP_021093402.1 | 4 - 5 | 808 | 88 | ||
XM_021237745.1 | XP_021093404.1 | 3 - 4 | 252 | 88 | ||
XM_021237746.1 | XP_021093405.1 | 4 - 5 | 410 | 88 | ||
XM_021237747.1 | XP_021093406.1 | 4 - 5 | 390 | 88 | ||
XM_021237748.1 | XP_021093407.1 | 3 - 4 | 317 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
