Assay Details
| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835669.3 | XP_004835726.1 | 3 - 4 | 631 | 71 | |
| XM_021263237.1 | XP_021118896.1 | 3 - 4 | 635 | 71 | ||
More Information
Set Membership: |
|

Timmdc1
Genomic Map