Assay Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840206.3 | XP_004840263.1 | 3 - 4 | 3173 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nyap1| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840206.3 | XP_004840263.1 | 3 - 4 | 3173 | 59 | |
Set Membership: |
|