Assay Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840203.3 | XP_004840260.1 | 4 - 5 | 3387 | 86 | |
| XM_004840204.3 | XP_004840261.1 | 5 - 6 | 2374 | 86 | ||
| XM_004840205.2 | XP_004840262.1 | 5 - 6 | 2314 | 86 | ||
| XM_004840206.3 | XP_004840263.1 | 4 - 5 | 3383 | 86 | ||
More Information
Set Membership: |
|

Nyap1
Genomic Map