Assay Details
| Gene Symbol: Steap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862394.2 | XP_004862451.1 | 3 - 4 | 746 | 70 | |
| XM_004862395.2 | XP_004862452.1 | 4 - 5 | 1147 | 70 | ||
| XM_004862396.2 | XP_004862453.1 | 3 - 4 | 963 | 70 | ||
| XM_004862397.2 | XP_004862454.1 | 3 - 4 | 751 | 70 | ||
More Information
Set Membership: |
|

Steap2
Genomic Map