Assay Details
| Gene Symbol: Ccdc113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843131.3 | XP_004843188.1 | 5 - 6 | 736 | 81 | |
| XM_004843132.3 | XP_004843189.1 | 4 - 5 | 575 | 81 | ||
More Information
Set Membership: |
|

Ccdc113
Genomic Map