Assay Details
| Gene Symbol: Tnfaip8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836159.3 | XP_004836216.1 | 1 - 2 | 189 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004836159.3 transcripts.
Set Membership: |
|

Tnfaip8
Genomic Map