Assay Details
| Gene Symbol: Sec61g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839842.3 | XP_004839899.1 | 3 - 4 | 328 | 91 | |
| XM_021254213.1 | XP_021109872.1 | 3 - 4 | 271 | 91 | ||
More Information
Set Membership: |
|

Sec61g
Genomic Map