Assay Details
| Gene Symbol: Mrpl54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865860.3 | XP_004865917.1 | 1 - 2 | 203 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl54| Gene Symbol: Mrpl54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865860.3 | XP_004865917.1 | 1 - 2 | 203 | 67 | |
Set Membership: |
|