Assay Details
| Gene Symbol: Manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862509.2 | XP_004862566.1 | 4 - 5 | 950 | 82 | |
| XM_021242669.1 | XP_021098328.1 | 4 - 5 | 869 | 82 | ||
More Information
Set Membership: |
|

Manea
Genomic Map