Assay Details
| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850506.3 | XP_004850563.1 | 11 - 12 | 1554 | 62 | |
| XM_013074427.2 | XP_012929881.1 | 8 - 9 | 1073 | 62 | ||
| XM_021263002.1 | XP_021118661.1 | 11 - 12 | 1439 | 62 | ||
More Information
Set Membership: |
|

Alpl
Genomic Map