Assay Details
| Gene Symbol: Arrdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848709.3 | XP_004848766.1 | 3 - 4 | 334 | 62 | |
| XM_013073912.2 | XP_012929366.1 | 3 - 4 | 337 | 62 | ||
More Information
Set Membership: |
|

Arrdc1
Genomic Map