Assay Details
| Gene Symbol: Ngf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853789.3 | XP_004853846.1 | 2 - 3 | 184 | 94 | |
| XM_021265739.1 | XP_021121398.1 | 2 - 3 | 233 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004853789.3 transcripts.
Set Membership: |
|

Ngf
Genomic Map