Assay Details
Gene Symbol: Fam162a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873600.3 | XP_004873657.1 | 2 - 3 | 268 | 57 | |
XM_021251260.1 | XP_021106919.1 | 2 - 3 | 268 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
