Assay Details
| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004833908.3 | XP_004833965.1 | 3 - 4 | 535 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btc| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004833908.3 | XP_004833965.1 | 3 - 4 | 535 | 54 | |
Set Membership: |
|