Assay Details
Gene Symbol: Gskip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004836992.2 | XP_004837049.1 | 1 - 2 | 113 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004836992.2 transcripts.
Set Membership: |
![]() ![]() |
