Assay Details
Gene Symbol: Gskip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004836992.2 | XP_004837049.1 | 2 - 3 | 369 | 64 | |
XM_021244572.1 | XP_021100231.1 | 2 - 3 | 1956 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
