Assay Details
| Gene Symbol: Zdhhc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843294.2 | XP_004843351.1 | 2 - 3 | 285 | 63 | |
| XM_004843295.3 | XP_004843352.1 | 2 - 3 | 281 | 63 | ||
More Information
Set Membership: |
|

Zdhhc8
Genomic Map