Assay Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867756.3 | XP_004867813.1 | 7 - 8 | 1169 | 62 | |
| XM_021246727.1 | XP_021102386.1 | 6 - 7 | 1060 | 62 | ||
| XM_021246728.1 | XP_021102387.1 | 5 - 6 | 746 | 62 | ||
More Information
Set Membership: |
|

Mat1a
Genomic Map