Assay Details
| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872890.3 | XP_004872947.2 | 2 - 3 | 292 | 86 | |
| XM_013068534.2 | XP_012923988.1 | 2 - 3 | 245 | 86 | ||
| XM_013068535.2 | XP_012923989.1 | 2 - 3 | 420 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013068534.2, XM_013068535.2 transcripts.
Set Membership: |
|

Tmem53
Genomic Map