Assay Details
| Gene Symbol: Chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844571.3 | XP_004844628.2 | 4 - 5 | 560 | 75 | |
| XM_021258196.1 | XP_021113855.1 | 4 - 5 | 506 | 75 | ||
More Information
Set Membership: |
|

Chmp3
Genomic Map