Assay Details
| Gene Symbol: Rab39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856509.2 | XP_004856566.1 | 1 - 2 | 339 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab39a| Gene Symbol: Rab39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856509.2 | XP_004856566.1 | 1 - 2 | 339 | 76 | |
Set Membership: |
|