Assay Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834773.3 | XP_004834830.1 | 1 - 2 | 330 | 66 | |
| XM_021255651.1 | XP_021111310.1 | 1 - 2 | 510 | 66 | ||
| XM_021255663.1 | XP_021111322.1 | 1 - 2 | 291 | 66 | ||
More Information
Set Membership: |
|

Ccdc50
Genomic Map