Assay Details
| Gene Symbol: Naif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261984.1 | XP_021117643.1 | 1 - 2 | 654 | 71 | |
| XM_021261985.1 | XP_021117644.1 | 1 - 2 | 668 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021261984.1, XM_021261985.1 transcripts.
Set Membership: |
|

Naif1
Genomic Map