Assay Details
| Gene Symbol: Naif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073569.2 | XP_012929023.1 | 1 - 2 | 1563 | 85 | |
| XM_021261984.1 | XP_021117643.1 | 2 - 3 | 1184 | 85 | ||
| XM_021261985.1 | XP_021117644.1 | 2 - 3 | 1198 | 85 | ||
| XM_021261986.1 | XP_021117645.1 | 2 - 3 | 1312 | 85 | ||
More Information
Set Membership: |
|

Naif1
Genomic Map