Assay Details
| Gene Symbol: Naif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073569.2 | XP_012929023.1 | 2 - 3 | 2077 | 61 | |
| XM_021261984.1 | XP_021117643.1 | 3 - 4 | 1698 | 61 | ||
| XM_021261986.1 | XP_021117645.1 | 3 - 4 | 1826 | 61 | ||
More Information
Set Membership: |
|

Naif1
Genomic Map