Assay Details
| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076845.2 | XP_012932299.1 | 11 - 12 | 1793 | 71 | |
| XM_013076846.2 | XP_012932300.1 | 10 - 11 | 1600 | 71 | ||
| XM_021238294.1 | XP_021093953.1 | 12 - 13 | 1829 | 71 | ||
| XM_021238295.1 | XP_021093954.1 | 10 - 11 | 1718 | 71 | ||
More Information
Set Membership: |
|

Pafah1b1
Genomic Map