Assay Details
| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845659.2 | XP_004845716.1 | 1 - 2 | 408 | 60 | |
| XM_004845660.2 | XP_004845717.1 | 1 - 2 | 409 | 60 | ||
More Information
Set Membership: |
|

Mfng
Genomic Map