Assay Details
Gene Symbol: Upf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872168.3 | XP_004872225.1 | 10 - 11 | 1407 | 66 | |
XM_004872169.3 | XP_004872226.1 | 9 - 10 | 1368 | 66 | ||
XM_021250129.1 | XP_021105788.1 | 9 - 10 | 1308 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
