Assay Details
| Gene Symbol: Fignl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839860.3 | XP_004839917.1 | 2 - 3 | 303 | 115 | |
| XM_021254229.1 | XP_021109888.1 | 2 - 3 | 628 | 115 | ||
| XM_021254230.1 | XP_021109889.1 | 2 - 3 | 389 | 115 | ||
| XM_021254231.1 | XP_021109890.1 | 2 - 3 | 153 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004839860.3, XM_021254229.1, XM_021254230.1, XM_021254231.1 transcripts.
Set Membership: |
|

Fignl1
Genomic Map