Assay Details
| Gene Symbol: Enosf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853179.3 | XP_004853236.1 | 1 - 2 | 217 | 53 | |
| XM_021265188.1 | XP_021120847.1 | 1 - 2 | 220 | 53 | ||
More Information
Set Membership: |
|

Enosf1
Genomic Map