Assay Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860600.3 | XP_004860657.1 | 9 - 10 | 903 | 62 | |
| XM_004860601.3 | XP_004860658.1 | 9 - 10 | 896 | 62 | ||
| XM_021241121.1 | XP_021096780.1 | 9 - 10 | 900 | 62 | ||
More Information
Set Membership: |
|

Tmem104
Genomic Map