Assay Details
| Gene Symbol: Art5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863445.3 | XP_004863502.1 | 2 - 3 | 474 | 112 | |
| XM_021243302.1 | XP_021098961.1 | 2 - 3 | 615 | 112 | ||
| XM_021243303.1 | XP_021098962.1 | 2 - 3 | 474 | 112 | ||
| XM_021243304.1 | XP_021098963.1 | 2 - 3 | 474 | 112 | ||
| XM_021243305.1 | XP_021098964.1 | 2 - 3 | 474 | 112 | ||
| XM_021243306.1 | XP_021098965.1 | 2 - 3 | 474 | 112 | ||
More Information
Set Membership: |
|

Art5
Genomic Map