Assay Details
| Gene Symbol: Wnt8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841947.2 | XP_004842004.1 | 4 - 5 | 504 | 67 | |
| XM_021255876.1 | XP_021111535.1 | 4 - 5 | 362 | 67 | ||
More Information
Set Membership: |
|

Wnt8a
Genomic Map