Assay Details
| Gene Symbol: Fam222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843607.3 | XP_004843664.1 | 2 - 3 | 310 | 69 | |
| XM_021257298.1 | XP_021112957.1 | 2 - 3 | 213 | 69 | ||
More Information
Set Membership: |
|

Fam222a
Genomic Map