Assay Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838436.3 | XP_004838493.1 | 5 - 6 | 519 | 61 | |
| XM_021252995.1 | XP_021108654.1 | 5 - 6 | 458 | 61 | ||
| XM_021252996.1 | XP_021108655.1 | 4 - 5 | 436 | 61 | ||
More Information
Set Membership: |
|

Dctn3
Genomic Map