Assay Details
| Gene Symbol: Dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841838.2 | XP_004841895.1 | 3 - 4 | 517 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc18| Gene Symbol: Dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841838.2 | XP_004841895.1 | 3 - 4 | 517 | 58 | |
Set Membership: |
|