Assay Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858930.3 | XP_004858987.1 | 8 - 9 | 837 | 58 | |
| XM_004858932.3 | XP_004858989.1 | 9 - 10 | 752 | 58 | ||
| XM_004858933.3 | XP_004858990.1 | 7 - 8 | 704 | 58 | ||
| XM_021239546.1 | XP_021095205.1 | 9 - 10 | 875 | 58 | ||
More Information
Set Membership: |
|

Lin37
Genomic Map