Assay Details
| Gene Symbol: Rnf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870329.3 | XP_004870386.1 | 6 - 7 | 1588 | 66 | |
| XM_021248665.1 | XP_021104324.1 | 5 - 6 | 1211 | 66 | ||
More Information
Set Membership: |
|

Rnf43
Genomic Map