Assay Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839547.2 | XP_004839604.1 | 6 - 7 | 905 | 70 | |
| XM_004839548.2 | XP_004839605.1 | 6 - 7 | 902 | 70 | ||
| XM_004839549.2 | XP_004839606.1 | 5 - 6 | 797 | 70 | ||
| XM_004839550.2 | XP_004839607.1 | 5 - 6 | 800 | 70 | ||
| XM_021254114.1 | XP_021109773.1 | 7 - 8 | 941 | 70 | ||
| XM_021254115.1 | XP_021109774.1 | 6 - 7 | 938 | 70 | ||
| XM_021254116.1 | XP_021109775.1 | 7 - 8 | 938 | 70 | ||
| XM_021254117.1 | XP_021109776.1 | 7 - 8 | 956 | 70 | ||
| XM_021254118.1 | XP_021109777.1 | 6 - 7 | 734 | 70 | ||
More Information
Set Membership: |
|

Fam221a
Genomic Map