Assay Details
| Gene Symbol: Fshr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839469.2 | XP_004839526.1 | 1 - 2 | 237 | 60 | |
| XM_021253847.1 | XP_021109506.1 | 1 - 2 | 237 | 60 | ||
More Information
Set Membership: |
|

Fshr
Genomic Map