Assay Details
| Gene Symbol: Mafg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860985.3 | XP_004861042.1 | 3 - 4 | 386 | 72 | |
| XM_013078241.2 | XP_012933695.1 | 2 - 3 | 255 | 72 | ||
More Information
Set Membership: |
|

Mafg
Genomic Map