Assay Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845030.3 | XP_004845087.1 | 6 - 7 | 991 | 63 | |
| XM_013072324.2 | XP_012927778.1 | 6 - 7 | 1073 | 63 | ||
| XM_021258525.1 | XP_021114184.1 | 5 - 6 | 1238 | 63 | ||
| XM_021258526.1 | XP_021114185.1 | 6 - 7 | 1133 | 63 | ||
More Information
Set Membership: |
|

Tmem19
Genomic Map