Assay Details
| Gene Symbol: Nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066313.2 | XP_012921767.1 | 3 - 4 | 346 | 59 | |
| XM_021245765.1 | XP_021101424.1 | 4 - 5 | 463 | 59 | ||
| XM_021245766.1 | XP_021101425.1 | 5 - 6 | 572 | 59 | ||
| XM_021245767.1 | XP_021101426.1 | 4 - 5 | 530 | 59 | ||
More Information
Set Membership: |
|

Nosip
Genomic Map