Assay Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845568.3 | XP_004845625.1 | 4 - 5 | 781 | 64 | |
| XM_004845570.3 | XP_004845627.1 | 4 - 5 | 768 | 64 | ||
More Information
Set Membership: |
|

Mief1
Genomic Map