Assay Details
| Gene Symbol: Kbtbd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875284.2 | XP_004875341.1 | 5 - 6 | 1743 | 64 | |
| XM_021252499.1 | XP_021108158.1 | 4 - 5 | 1685 | 64 | ||
More Information
Set Membership: |
|

Kbtbd12
Genomic Map