Assay Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257015.1 | XP_021112674.1 | 1 - 2 | 167 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021257015.1 transcripts.
Set Membership: |
|

Fam192a
Genomic Map